Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.0765 Inch Diameter, 2 Inch Long
Catalog No. 5000530260
Encompass
This Vermont Gage Class X minus pin gage features a precise 0.0765 inch diameter and a length of 2 inches. It is manufactured from durable tool steel and boasts a 4 micro coated finish. This pin gage is ideal for the calibration of measuring equipment or for directly gaging parts and comes with a Certificate of Accuracy that provides NIST Traceability.
  • Precise 0.0765 inch diameter
  • 2 inch length
  • Durable tool steel material
  • 4 micro coated finish
  • Ideal for calibration of measuring equipment
  • Suitable for directly gaging parts
  • Includes a certificate of accuracy with NIST traceability
  • Tolerance of -0.00004 decimal inches
Catalog No. 50-005-30260 Supplier MSC Supplier No. 88807656
This Vermont Gage Class X minus pin gage features a precise 0.0765 inch diameter and a length of 2 inches. It is manufactured from durable tool steel and boasts a 4 micro coated finish. This pin gage is ideal for the calibration of measuring equipment or for directly gaging parts and comes with a Certificate of Accuracy that provides NIST Traceability.
  • Precise 0.0765 inch diameter
  • 2 inch length
  • Durable tool steel material
  • 4 micro coated finish
  • Ideal for calibration of measuring equipment
  • Suitable for directly gaging parts
  • Includes a certificate of accuracy with NIST traceability
  • Tolerance of -0.00004 decimal inches