Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.0785 in, 2 in
Catalog No. 5000530940
Encompass
This Class X Minus Pin Gage is designed for precise measurement applications. It features a 0.0785 inch diameter, a 2 inch length, and a 4 Micro Coated finish, making it suitable for calibrating measuring equipment or directly gaging parts.
  • 4 Micro Coated finish
  • Includes certificate of accuracy with NIST traceability
  • Suitable for calibration of measuring equipment or to gage parts directly
  • Made from tool steel
Catalog No. 50-005-30940 Supplier MSC Supplier No. 88807854
This Class X Minus Pin Gage is designed for precise measurement applications. It features a 0.0785 inch diameter, a 2 inch length, and a 4 Micro Coated finish, making it suitable for calibrating measuring equipment or directly gaging parts.
  • 4 Micro Coated finish
  • Includes certificate of accuracy with NIST traceability
  • Suitable for calibration of measuring equipment or to gage parts directly
  • Made from tool steel