Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.0787 in, 2 in
Catalog No. 5000534394
Encompass
The Vermont Gage Class X Minus Pin Gage is designed for precision measurement, ideal for the calibration of measuring equipment or direct part gaging. It features a 0.0787 in diameter and a 2 in length, crafted from durable tool steel.
  • 0.0787 in diameter
  • -0.00004 in tolerance
  • Tool steel material
  • Includes certificate of accuracy with NIST traceability
  • Suitable for calibration of measuring equipment or gaging parts directly
  • 4 micro finish
Catalog No. 50-005-34394 Supplier MSC Supplier No. 88126040
The Vermont Gage Class X Minus Pin Gage is designed for precision measurement, ideal for the calibration of measuring equipment or direct part gaging. It features a 0.0787 in diameter and a 2 in length, crafted from durable tool steel.
  • 0.0787 in diameter
  • -0.00004 in tolerance
  • Tool steel material
  • Includes certificate of accuracy with NIST traceability
  • Suitable for calibration of measuring equipment or gaging parts directly
  • 4 micro finish