Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.0935 in Diameter, 2 in Length, Tool Steel
Catalog No. 5000531153
Encompass
This Class X Minus Pin Gage features a precise 0.0935 in diameter and a 2 in length, crafted from durable tool steel with a 4 micro finish. It is designed for accurate calibration of measuring equipment or for direct part gaging, ensuring reliable measurements in various applications.
  • Diameter of measurement: 0.0935 in
  • Tolerance: -0.00004 in
  • Made from tool steel
  • Designed for calibration of measuring equipment or direct part gaging
  • Includes certificate of accuracy with NIST traceability
  • Length: 2 in
  • Features a 4 micro finish
Catalog No. 50-005-31153 Supplier MSC Supplier No. 88809355
This Class X Minus Pin Gage features a precise 0.0935 in diameter and a 2 in length, crafted from durable tool steel with a 4 micro finish. It is designed for accurate calibration of measuring equipment or for direct part gaging, ensuring reliable measurements in various applications.
  • Diameter of measurement: 0.0935 in
  • Tolerance: -0.00004 in
  • Made from tool steel
  • Designed for calibration of measuring equipment or direct part gaging
  • Includes certificate of accuracy with NIST traceability
  • Length: 2 in
  • Features a 4 micro finish