Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.0960 in, 2 in Long
Catalog No. 5000527470
Encompass
This Vermont Gage Class X Minus Pin Gage is designed for precise measurement and calibration tasks. Featuring a 0.0960 in diameter and 2 in length, it is made from durable tool steel and has a 4 micro coated finish for enhanced performance. It is suitable for calibrating measuring equipment or directly gaging parts, and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for precise measurement and calibration
  • 0.0960 in diameter
  • 2 in long
  • Constructed from tool steel
  • Features a 4 micro coated finish
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured in the United States
Catalog No. 50-005-27470 Supplier MSC Supplier No. 88809603
This Vermont Gage Class X Minus Pin Gage is designed for precise measurement and calibration tasks. Featuring a 0.0960 in diameter and 2 in length, it is made from durable tool steel and has a 4 micro coated finish for enhanced performance. It is suitable for calibrating measuring equipment or directly gaging parts, and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for precise measurement and calibration
  • 0.0960 in diameter
  • 2 in long
  • Constructed from tool steel
  • Features a 4 micro coated finish
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured in the United States