Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.1247 in Diameter, 2 in Length, Tool Steel
Catalog No. 5000510494
Encompass
This Class X Minus Pin Gage is designed for precise measurement and calibration tasks. With a diameter of 0.1247 inch and a length of 2 inch, this tool steel gage provides accuracy for verifying measurements or directly gaging parts. It includes a Certificate of Accuracy with NIST traceability, ensuring reliable performance.
  • Made from durable tool steel.
  • Designed for calibrating measuring equipment or direct part gaging.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 4 micro coating/finish for enhanced performance.
  • Manufactured to Class X tolerance standards for precise measurements.
Catalog No. 50-005-10494 Supplier MSC Supplier No. 71044044
This Class X Minus Pin Gage is designed for precise measurement and calibration tasks. With a diameter of 0.1247 inch and a length of 2 inch, this tool steel gage provides accuracy for verifying measurements or directly gaging parts. It includes a Certificate of Accuracy with NIST traceability, ensuring reliable performance.
  • Made from durable tool steel.
  • Designed for calibrating measuring equipment or direct part gaging.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 4 micro coating/finish for enhanced performance.
  • Manufactured to Class X tolerance standards for precise measurements.