Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.1300 in Diameter, 2 in Length, Tool Steel
Catalog No. 5000527562
Encompass
This Class X Minus Pin Gage is designed for the accurate calibration of measuring equipment or for direct part gaging. Constructed from durable tool steel, it features a precise 0.1300 in diameter and a 2 in length, ensuring reliability for metrology applications.
  • Accurate, cost-effective measurement standard.
  • Suitable for calibration of measuring equipment or direct part gaging.
  • Constructed from tool steel for durability.
  • Features a 4 microinch finish or better.
  • Round within 1/2 the diameter tolerance.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-27562 Supplier MSC Supplier No. 88813001
This Class X Minus Pin Gage is designed for the accurate calibration of measuring equipment or for direct part gaging. Constructed from durable tool steel, it features a precise 0.1300 in diameter and a 2 in length, ensuring reliability for metrology applications.
  • Accurate, cost-effective measurement standard.
  • Suitable for calibration of measuring equipment or direct part gaging.
  • Constructed from tool steel for durability.
  • Features a 4 microinch finish or better.
  • Round within 1/2 the diameter tolerance.
  • Includes a certificate of accuracy with NIST traceability.