Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.1915 in, 2 in, Tool Steel
Catalog No. 5000535390
Encompass
This class X minus pin gage is designed for precise measurement and calibration tasks. With a diameter of 0.1915 inches and a length of 2 inches, it is constructed from durable tool steel and features a 4 micro coating. It is suitable for calibrating measuring equipment or directly gaging parts.
  • Diameter of 0.1915 in
  • Tolerance of -0.00004 in
  • Made from tool steel
  • Includes certificate of accuracy with NIST traceability
  • Length of 2 in
  • Features 4 micro coating
  • Suitable for calibration of measuring equipment or gaging parts directly
Catalog No. 50-005-35390 Supplier MSC Supplier No. 88819156
This class X minus pin gage is designed for precise measurement and calibration tasks. With a diameter of 0.1915 inches and a length of 2 inches, it is constructed from durable tool steel and features a 4 micro coating. It is suitable for calibrating measuring equipment or directly gaging parts.
  • Diameter of 0.1915 in
  • Tolerance of -0.00004 in
  • Made from tool steel
  • Includes certificate of accuracy with NIST traceability
  • Length of 2 in
  • Features 4 micro coating
  • Suitable for calibration of measuring equipment or gaging parts directly