Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.1960 in, 2 in, 4 Micro Coated, Tool Steel
Catalog No. 5000531708
Encompass
This Class X Minus Pin Gage is designed for the calibration of measuring equipment or for direct part gaging. It features a measurement diameter of 0.1960 in and a length of 2 in. Constructed from durable tool steel, it also includes a 4 Micro coating/finish and has a tolerance of -0.00004 in. Each gage comes with a Certificate of Accuracy with NIST Traceability.
  • Designed for calibration of measuring equipment
  • Can be used for direct part gaging
  • Features a measurement diameter of 0.1960 in
  • Has a length of 2 in
  • Constructed from durable tool steel
  • Includes a 4 Micro coating/finish
  • Tolerance of -0.00004 in
  • Comes with a Certificate of Accuracy with NIST Traceability
  • Made in the United States
Catalog No. 50-005-31708 Supplier MSC Supplier No. 88819602
This Class X Minus Pin Gage is designed for the calibration of measuring equipment or for direct part gaging. It features a measurement diameter of 0.1960 in and a length of 2 in. Constructed from durable tool steel, it also includes a 4 Micro coating/finish and has a tolerance of -0.00004 in. Each gage comes with a Certificate of Accuracy with NIST Traceability.
  • Designed for calibration of measuring equipment
  • Can be used for direct part gaging
  • Features a measurement diameter of 0.1960 in
  • Has a length of 2 in
  • Constructed from durable tool steel
  • Includes a 4 Micro coating/finish
  • Tolerance of -0.00004 in
  • Comes with a Certificate of Accuracy with NIST Traceability
  • Made in the United States