Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.2150 in, 2 in, 4 Micro
Catalog No. 5000536749
Encompass
This Class X Minus Pin Gage has a diameter of 0.2150 in and a length of 2 in. It features a 4 Micro coating and is made of tool steel. The gage includes a Certificate of Accuracy with NIST Traceability and is designed for the calibration of measuring equipment or for directly gaging parts.
  • 4 Micro coating
  • Certificate of accuracy with NIST traceability included
  • Made from tool steel
  • Ideal for calibration of measuring equipment or direct gaging of parts
Catalog No. 50-005-36749 Supplier MSC Supplier No. 88821509
This Class X Minus Pin Gage has a diameter of 0.2150 in and a length of 2 in. It features a 4 Micro coating and is made of tool steel. The gage includes a Certificate of Accuracy with NIST Traceability and is designed for the calibration of measuring equipment or for directly gaging parts.
  • 4 Micro coating
  • Certificate of accuracy with NIST traceability included
  • Made from tool steel
  • Ideal for calibration of measuring equipment or direct gaging of parts