Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.2490 in, 2 in, 4 Micro
Catalog No. 5000510445
Encompass
The Vermont Gage Class X Minus Pin Gage is designed for precise measurement and calibration tasks. Made from tool steel with a 4 Micro coating, this 0.2490 inch diameter and 2 inch long gage ensures accuracy. It is suitable for calibrating measuring equipment or directly gaging parts and includes a Certificate of Accuracy with NIST Traceability.
  • Made from durable tool steel.
  • Features a 4 Micro coating for enhanced precision.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for calibrating measuring equipment.
  • Can be used to gage parts directly.
  • Manufactured in the United States.
Catalog No. 50-005-10445 Supplier MSC Supplier No. 88824909
The Vermont Gage Class X Minus Pin Gage is designed for precise measurement and calibration tasks. Made from tool steel with a 4 Micro coating, this 0.2490 inch diameter and 2 inch long gage ensures accuracy. It is suitable for calibrating measuring equipment or directly gaging parts and includes a Certificate of Accuracy with NIST Traceability.
  • Made from durable tool steel.
  • Features a 4 Micro coating for enhanced precision.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for calibrating measuring equipment.
  • Can be used to gage parts directly.
  • Manufactured in the United States.