Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.2980 in, 2 in, Tool Steel
Catalog No. 5000527468
Encompass
This Vermont Gage Class X Minus Pin Gage is designed for precise measurement applications, serving as an accurate and cost-effective standard for calibrating equipment or direct part gaging. Crafted from durable tool steel, it features a 0.2980 in diameter and a 2 in length with a 4 microinch finish. Each gage includes a Certificate of Accuracy.
  • Used as masters for calibrating equipment or for direct part gaging.
  • Tool steel construction with 60/65 Rc hardness.
  • 2 in long.
  • 4 microinch or better finish.
  • Includes a certificate of accuracy.
  • Features minus tolerance for precise measurement.
Catalog No. 50-005-27468 Supplier MSC Supplier No. 88829809
This Vermont Gage Class X Minus Pin Gage is designed for precise measurement applications, serving as an accurate and cost-effective standard for calibrating equipment or direct part gaging. Crafted from durable tool steel, it features a 0.2980 in diameter and a 2 in length with a 4 microinch finish. Each gage includes a Certificate of Accuracy.
  • Used as masters for calibrating equipment or for direct part gaging.
  • Tool steel construction with 60/65 Rc hardness.
  • 2 in long.
  • 4 microinch or better finish.
  • Includes a certificate of accuracy.
  • Features minus tolerance for precise measurement.