Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.3170 Inch Diameter, 2 Inch Length, Tool Steel
Catalog No. 5000527469
Encompass
This Vermont Gage Class X Minus Pin Gage is designed for precise measurement and calibration tasks. Crafted from durable tool steel, it features a 0.3170 inch diameter and a 2 inch length. Each gage is finished to 4 micro inches and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable and accurate performance for quality control and inspection applications.
  • Crafted from durable tool steel.
  • Features a precise 0.3170 inch diameter.
  • Measures 2 inches in length.
  • Finished to 4 micro inches for smooth surface.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Ideal for calibration of measuring equipment or gaging parts directly.
Catalog No. 50-005-27469 Supplier MSC Supplier No. 88831706
This Vermont Gage Class X Minus Pin Gage is designed for precise measurement and calibration tasks. Crafted from durable tool steel, it features a 0.3170 inch diameter and a 2 inch length. Each gage is finished to 4 micro inches and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable and accurate performance for quality control and inspection applications.
  • Crafted from durable tool steel.
  • Features a precise 0.3170 inch diameter.
  • Measures 2 inches in length.
  • Finished to 4 micro inches for smooth surface.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Ideal for calibration of measuring equipment or gaging parts directly.