Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.3980 in, 2 in
Catalog No. 5000530743
Encompass
This Class X Minus Pin Gage features a 0.3980 in diameter and a 2 in length, with a 4 Micro finish. It is constructed from tool steel and is suitable for the calibration of measuring equipment or for direct gaging of parts. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • 0.3980 in diameter
  • -0.00004 in tolerance
  • Tool steel material
  • 2 in length
  • 4 Micro finish
  • Suitable for calibration of measuring equipment or direct gaging of parts
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-30743 Supplier MSC Supplier No. 88839808
This Class X Minus Pin Gage features a 0.3980 in diameter and a 2 in length, with a 4 Micro finish. It is constructed from tool steel and is suitable for the calibration of measuring equipment or for direct gaging of parts. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • 0.3980 in diameter
  • -0.00004 in tolerance
  • Tool steel material
  • 2 in length
  • 4 Micro finish
  • Suitable for calibration of measuring equipment or direct gaging of parts
  • Includes certificate of accuracy with NIST traceability