Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.5200 in Dia, 2 in Long, 4 Micro Coated
Catalog No. 5000530502
Encompass
This Class X Minus Pin Gage is manufactured by Vermont Gage. It features a precise 0.5200 inch diameter and a 2 inch length, with a 4 Micro coating for enhanced durability and accuracy. This gage is ideal for the calibration of measuring equipment or for direct part gaging.
  • Constructed from durable tool steel
  • Features a 4 Micro coating
  • Includes a Certificate of Accuracy with NIST Traceability
  • Suitable for calibrating measuring equipment or directly gaging parts
Catalog No. 50-005-30502 Supplier MSC Supplier No. 88852009
This Class X Minus Pin Gage is manufactured by Vermont Gage. It features a precise 0.5200 inch diameter and a 2 inch length, with a 4 Micro coating for enhanced durability and accuracy. This gage is ideal for the calibration of measuring equipment or for direct part gaging.
  • Constructed from durable tool steel
  • Features a 4 Micro coating
  • Includes a Certificate of Accuracy with NIST Traceability
  • Suitable for calibrating measuring equipment or directly gaging parts