Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.8740 in Diameter, 2 in Long, Tool Steel
Catalog No. 5000528858
Encompass
This Class X Minus Pin Gage is designed for the precise calibration of measuring equipment or for directly gaging parts. It features a 0.8740 inch diameter and a 2 inch length, manufactured from durable tool steel. The gage is finished with a 4 micro coating and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable and accurate measurements.
  • Designed for accurate calibration of measuring equipment
  • Suitable for directly gaging parts
  • Constructed from durable tool steel
  • Features a 4 micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Offers precise measurements for demanding applications
Catalog No. 50-005-28858 Supplier MSC Supplier No. 88887401
This Class X Minus Pin Gage is designed for the precise calibration of measuring equipment or for directly gaging parts. It features a 0.8740 inch diameter and a 2 inch length, manufactured from durable tool steel. The gage is finished with a 4 micro coating and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable and accurate measurements.
  • Designed for accurate calibration of measuring equipment
  • Suitable for directly gaging parts
  • Constructed from durable tool steel
  • Features a 4 micro finish
  • Includes a certificate of accuracy with NIST traceability
  • Offers precise measurements for demanding applications