Learn More
MSC Vermont Gage Class X Plug Gage, 0.4350 in Dia, 2 in Long, Tool Steel
Catalog No. 5000528860
Encompass
This plug gage is designed for precise measurement and calibration in both shop floor and metrology laboratory settings. It provides accurate, cost-effective measurement standards for ensuring component quality and equipment accuracy.
  • Class X tolerance with a positive deviation of +0.00004 in and no negative deviation for sizes up to 0.8250 in.
  • Constructed from cold stabilized tool steel (60/65 Rc) for durability.
  • Features a fine 4 microinch finish or better.
  • NIST traceable for reliable measurement standards.
  • Includes a certificate of accuracy.
  • Measures 2 in long.
  • Has a diameter of 0.4350 in.
  • Roundness is maintained within 1/2 of the specified tolerance.
Catalog No. 50-005-28860 Supplier MSC Supplier No. 88743505
This plug gage is designed for precise measurement and calibration in both shop floor and metrology laboratory settings. It provides accurate, cost-effective measurement standards for ensuring component quality and equipment accuracy.
  • Class X tolerance with a positive deviation of +0.00004 in and no negative deviation for sizes up to 0.8250 in.
  • Constructed from cold stabilized tool steel (60/65 Rc) for durability.
  • Features a fine 4 microinch finish or better.
  • NIST traceable for reliable measurement standards.
  • Includes a certificate of accuracy.
  • Measures 2 in long.
  • Has a diameter of 0.4350 in.
  • Roundness is maintained within 1/2 of the specified tolerance.