Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.0200 in Diameter, 2 in Length, 4 Micro Finish
Catalog No. 5000530925
Encompass
This Class X Plus pin gage serves as a precise measurement standard for equipment calibration or direct part gaging in both shop floor and metrology laboratory applications. It features a 0.0200 inch diameter, 2 inch length, and is crafted from durable tool steel with a fine finish.
  • Designed for accurate and cost-effective measurement
  • Ideal for equipment calibration or direct part gaging
  • Constructed from tool steel for durability
  • Features a 4 microinch finish or better
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to a tolerance of +0.00004 inch
Catalog No. 50-005-30925 Supplier MSC Supplier No. 88702006
This Class X Plus pin gage serves as a precise measurement standard for equipment calibration or direct part gaging in both shop floor and metrology laboratory applications. It features a 0.0200 inch diameter, 2 inch length, and is crafted from durable tool steel with a fine finish.
  • Designed for accurate and cost-effective measurement
  • Ideal for equipment calibration or direct part gaging
  • Constructed from tool steel for durability
  • Features a 4 microinch finish or better
  • Includes a certificate of accuracy with NIST traceability
  • Manufactured to a tolerance of +0.00004 inch