Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.0460 in, 2 in
Catalog No. 5000534560
Encompass
This pin gage is designed for precise measurement and calibration applications. Made from durable tool steel, it features a 0.0460 inch diameter and a 2 inch length, ensuring accuracy for critical tasks.
  • Diameter of 0.0460 in
  • Length of 2 in
  • Tolerance of 0.00004 in
  • Constructed from tool steel
  • Features a 4 micro coating and finish
  • Ideal for calibration of measuring equipment or gaging parts directly
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-34560 Supplier MSC Supplier No. 88704606
This pin gage is designed for precise measurement and calibration applications. Made from durable tool steel, it features a 0.0460 inch diameter and a 2 inch length, ensuring accuracy for critical tasks.
  • Diameter of 0.0460 in
  • Length of 2 in
  • Tolerance of 0.00004 in
  • Constructed from tool steel
  • Features a 4 micro coating and finish
  • Ideal for calibration of measuring equipment or gaging parts directly
  • Includes certificate of accuracy with NIST traceability