Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.1320 in Diameter, 2 in Long, Tool Steel
Catalog No. 5000534507
Encompass
The Vermont Gage Class X Plus Pin Gage is a precise measurement standard designed for calibrating measuring equipment or direct part gaging. It features a 0.1320 inch diameter and a 2 inch length, crafted from durable tool steel with a 4 microinch finish. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Accurate and cost-effective measurement standard.
  • Suitable for both the shop floor and metrology laboratory.
  • Diameter of 0.1320 "
  • Length of 2 "
  • Made from durable tool steel.
  • Features a 4 microinch finish or better.
  • Certificate of accuracy with NIST traceability included.
Catalog No. 50-005-34507 Supplier MSC Supplier No. 88713201
The Vermont Gage Class X Plus Pin Gage is a precise measurement standard designed for calibrating measuring equipment or direct part gaging. It features a 0.1320 inch diameter and a 2 inch length, crafted from durable tool steel with a 4 microinch finish. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Accurate and cost-effective measurement standard.
  • Suitable for both the shop floor and metrology laboratory.
  • Diameter of 0.1320 "
  • Length of 2 "
  • Made from durable tool steel.
  • Features a 4 microinch finish or better.
  • Certificate of accuracy with NIST traceability included.