Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.1360" Dia, 2" Long, 4 Micro Coated
Catalog No. 5000534533
Encompass
This Class X Plus Pin Gage features a diameter of 0.1360" and a length of 2". It is designed with a 4 Micro Coated finish, making it suitable for precise calibration of measuring equipment or directly gaging parts.
  • Class X Plus Pin Gage
  • 0.1360" Diameter
  • 2" Length
  • 4 Micro Coated finish
  • Includes certificate of accuracy with NIST traceability
  • Suitable for calibration of measuring equipment or directly gaging parts
Catalog No. 50-005-34533 Supplier MSC Supplier No. 88713607
This Class X Plus Pin Gage features a diameter of 0.1360" and a length of 2". It is designed with a 4 Micro Coated finish, making it suitable for precise calibration of measuring equipment or directly gaging parts.
  • Class X Plus Pin Gage
  • 0.1360" Diameter
  • 2" Length
  • 4 Micro Coated finish
  • Includes certificate of accuracy with NIST traceability
  • Suitable for calibration of measuring equipment or directly gaging parts