Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.1520 in, 2 in
Catalog No. 5000531220
Encompass
This pin gage is a precision measuring tool designed for accurate calibration and gaging tasks. Manufactured from durable tool steel, it features a Class X Plus tolerance, ensuring high accuracy for demanding applications. With a diameter of 0.1520 inches and a length of 2 inches, it is versatile for various measurement requirements.
  • Class X Plus tolerance for high precision measurements
  • Diameter of 0.1520 in
  • Length of 2 in
  • Constructed from tool steel for enhanced durability
  • Features a 4 micro coating/finish
  • Ideal for the calibration of measuring equipment
  • Can be used for direct part gaging
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-31220 Supplier MSC Supplier No. 88715206
This pin gage is a precision measuring tool designed for accurate calibration and gaging tasks. Manufactured from durable tool steel, it features a Class X Plus tolerance, ensuring high accuracy for demanding applications. With a diameter of 0.1520 inches and a length of 2 inches, it is versatile for various measurement requirements.
  • Class X Plus tolerance for high precision measurements
  • Diameter of 0.1520 in
  • Length of 2 in
  • Constructed from tool steel for enhanced durability
  • Features a 4 micro coating/finish
  • Ideal for the calibration of measuring equipment
  • Can be used for direct part gaging
  • Includes a certificate of accuracy with NIST traceability