Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.1920 in, 2 in
Catalog No. 5000531608
Encompass
This Class X Plus Pin Gage is designed for precise measurement and calibration tasks. Manufactured from durable tool steel, it features a 0.1920 inch diameter and a two inch length, along with a 4 micro finish for enhanced accuracy.
  • Manufactured from tool steel for durability.
  • Features a 0.1920 in diameter for precise measurements.
  • Measures 2 in length, suitable for various applications.
  • Includes a 4 micro finish for enhanced surface quality.
  • Ideal for calibration of measuring equipment or direct part gaging.
  • Supplied with a certificate of accuracy with NIST traceability.
Catalog No. 50-005-31608 Supplier MSC Supplier No. 88719208
This Class X Plus Pin Gage is designed for precise measurement and calibration tasks. Manufactured from durable tool steel, it features a 0.1920 inch diameter and a two inch length, along with a 4 micro finish for enhanced accuracy.
  • Manufactured from tool steel for durability.
  • Features a 0.1920 in diameter for precise measurements.
  • Measures 2 in length, suitable for various applications.
  • Includes a 4 micro finish for enhanced surface quality.
  • Ideal for calibration of measuring equipment or direct part gaging.
  • Supplied with a certificate of accuracy with NIST traceability.