Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.1930 in, 2 in, Tool Steel
Catalog No. 5000531150
Encompass
This Class X Plus Pin Gage is designed for precision measurement applications. Made from durable tool steel, it features a 0.1930 in diameter and a 2 in length, suitable for calibrating measuring equipment or directly gaging parts.
  • Features a diameter of 0.1930 in.
  • Provides a tolerance of 0.00004 in.
  • Constructed from tool steel for durability.
  • Ideal for calibration of measuring equipment or direct part gaging.
  • Includes a certificate of accuracy with NIST traceability.
  • Measures 2 in in length.
  • Finished with a 4 micro coating.
  • Manufactured in the United States.
Catalog No. 50-005-31150 Supplier MSC Supplier No. 88719307
This Class X Plus Pin Gage is designed for precision measurement applications. Made from durable tool steel, it features a 0.1930 in diameter and a 2 in length, suitable for calibrating measuring equipment or directly gaging parts.
  • Features a diameter of 0.1930 in.
  • Provides a tolerance of 0.00004 in.
  • Constructed from tool steel for durability.
  • Ideal for calibration of measuring equipment or direct part gaging.
  • Includes a certificate of accuracy with NIST traceability.
  • Measures 2 in in length.
  • Finished with a 4 micro coating.
  • Manufactured in the United States.