Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.2090 in, 2 in, Tool Steel
Catalog No. 5000531327
Encompass
This Class X Plus pin gage is precisely crafted with a 0.2090 inch diameter and a 2 inch length, made from durable tool steel. It features a 4 micro coating and is ideal for the calibration of measuring equipment or for direct part gaging applications.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from tool steel for durability.
Catalog No. 50-005-31327 Supplier MSC Supplier No. 88720909
This Class X Plus pin gage is precisely crafted with a 0.2090 inch diameter and a 2 inch length, made from durable tool steel. It features a 4 micro coating and is ideal for the calibration of measuring equipment or for direct part gaging applications.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from tool steel for durability.