Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.2310 in, 2 in, Tool Steel
Catalog No. 5000531149
Encompass
The Vermont Gage Class X Plus Pin Gage is a precision measuring tool designed for calibrating equipment or directly gaging parts. Made from durable tool steel, it features a 4 micro coating and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable and accurate measurements.
  • Designed for calibration of measuring equipment or direct gaging of parts
  • Made from durable tool steel
  • Features a 4 micro coating
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-31149 Supplier MSC Supplier No. 88723101
The Vermont Gage Class X Plus Pin Gage is a precision measuring tool designed for calibrating equipment or directly gaging parts. Made from durable tool steel, it features a 4 micro coating and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable and accurate measurements.
  • Designed for calibration of measuring equipment or direct gaging of parts
  • Made from durable tool steel
  • Features a 4 micro coating
  • Includes a certificate of accuracy with NIST traceability