Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.3150 in, 2 in, Tool Steel
Catalog No. 5000510991
Encompass
This precision Class X Plus pin gage is designed for calibrating measuring equipment or directly gaging parts. It offers high accuracy with a 4 microinch finish and includes a Certificate of Accuracy with NIST Traceability, providing reliable measurement standards for various applications.
  • Tool steel construction for durability
  • 2 in length
  • 4 microinch finish or better
  • Certificate of accuracy with NIST traceability included
  • Suitable for calibrating equipment or direct part gaging
  • Roundness within 1/2 the diameter tolerance
Catalog No. 50-005-10991 Supplier MSC Supplier No. 88731500
This precision Class X Plus pin gage is designed for calibrating measuring equipment or directly gaging parts. It offers high accuracy with a 4 microinch finish and includes a Certificate of Accuracy with NIST Traceability, providing reliable measurement standards for various applications.
  • Tool steel construction for durability
  • 2 in length
  • 4 microinch finish or better
  • Certificate of accuracy with NIST traceability included
  • Suitable for calibrating equipment or direct part gaging
  • Roundness within 1/2 the diameter tolerance