Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.3230 in Diameter, 2 in Length, 4 Micro Coated
Catalog No. 5000528859
Encompass
This pin gage features a 0.3230 in diameter and a 2 in length, with a 4 Micro coating. It is constructed from durable tool steel and is designed for the calibration of measuring equipment or for direct gaging of parts. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring precision and reliability. Manufactured in the United States, this pin gage meets a tolerance of 0.00004 decimal inches.
  • 4 Micro coated
  • Includes certificate of accuracy with NIST traceability
  • Designed for calibration of measuring equipment or to gage parts directly
  • Constructed from tool steel
  • Meets a tolerance of 0.00004 decimal inches
  • Manufactured in the United States
Catalog No. 50-005-28859 Supplier MSC Supplier No. 88732300
This pin gage features a 0.3230 in diameter and a 2 in length, with a 4 Micro coating. It is constructed from durable tool steel and is designed for the calibration of measuring equipment or for direct gaging of parts. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring precision and reliability. Manufactured in the United States, this pin gage meets a tolerance of 0.00004 decimal inches.
  • 4 Micro coated
  • Includes certificate of accuracy with NIST traceability
  • Designed for calibration of measuring equipment or to gage parts directly
  • Constructed from tool steel
  • Meets a tolerance of 0.00004 decimal inches
  • Manufactured in the United States