Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.3580 in, 2 in Long, Tool Steel
Catalog No. 5000535374
Encompass
These Class X Plus Pin Gages are engineered for precise measurement and calibration. They are designed to be used as masters for calibrating measuring equipment or for direct gaging of parts. Providing accurate and cost-effective measurement standards, they are suitable for both shop floor and metrology laboratory environments.
  • Class X tolerance for high accuracy
  • NIST traceable, ensuring reliability
  • Constructed from cold stabilized tool steel for durability
  • Features a 4 microinch finish or better
  • Roundness maintained within 1/2 of the stated tolerance
  • Includes a certificate of accuracy
  • Go (Plus) or NoGo (Minus) tolerance available for various applications
  • Available in various sizes with 0.0001 inch increments
Catalog No. 50-005-35374 Supplier MSC Supplier No. 88735808
These Class X Plus Pin Gages are engineered for precise measurement and calibration. They are designed to be used as masters for calibrating measuring equipment or for direct gaging of parts. Providing accurate and cost-effective measurement standards, they are suitable for both shop floor and metrology laboratory environments.
  • Class X tolerance for high accuracy
  • NIST traceable, ensuring reliability
  • Constructed from cold stabilized tool steel for durability
  • Features a 4 microinch finish or better
  • Roundness maintained within 1/2 of the stated tolerance
  • Includes a certificate of accuracy
  • Go (Plus) or NoGo (Minus) tolerance available for various applications
  • Available in various sizes with 0.0001 inch increments