Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.3605 in Dia, 2 in Long, Tool Steel
Catalog No. 5000529035
Encompass
This Class X Plus Pin Gage features a precise 0.3605 inch diameter and a 2 inch length, crafted from durable tool steel. It is designed for calibrating measuring equipment or directly gaging parts.
  • Designed for calibration of measuring equipment or direct part gaging.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from tool steel for durability.
  • Provides a precise 0.3605 inch diameter measurement.
  • Features a 0.00004 inch tolerance.
  • Has a 4 micro coating/finish.
Catalog No. 50-005-29035 Supplier MSC Supplier No. 88736053
This Class X Plus Pin Gage features a precise 0.3605 inch diameter and a 2 inch length, crafted from durable tool steel. It is designed for calibrating measuring equipment or directly gaging parts.
  • Designed for calibration of measuring equipment or direct part gaging.
  • Includes a certificate of accuracy with NIST traceability.
  • Constructed from tool steel for durability.
  • Provides a precise 0.3605 inch diameter measurement.
  • Features a 0.00004 inch tolerance.
  • Has a 4 micro coating/finish.