Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.3755 in Diameter, 2 in Length, 4 Micro Finish
Catalog No. 5000531363
Encompass
This Class X Plus Pin Gage is designed for precision measurement applications, serving as a master for calibrating equipment or directly gaging parts. It provides accurate and cost-effective measurement standards suitable for both shop floor and metrology laboratories.
  • Designed for calibration of measuring equipment or direct part gaging.
  • Manufactured from durable tool steel with a 60-65 Rockwell C hardness.
  • Features a 0.3755 in diameter with a tight tolerance of 0.00004 in.
  • 2 in length for versatile application.
  • Smooth 4 microinch finish or better for precise measurements.
  • Certificate of accuracy with NIST traceability included.
Catalog No. 50-005-31363 Supplier MSC Supplier No. 88737556
This Class X Plus Pin Gage is designed for precision measurement applications, serving as a master for calibrating equipment or directly gaging parts. It provides accurate and cost-effective measurement standards suitable for both shop floor and metrology laboratories.
  • Designed for calibration of measuring equipment or direct part gaging.
  • Manufactured from durable tool steel with a 60-65 Rockwell C hardness.
  • Features a 0.3755 in diameter with a tight tolerance of 0.00004 in.
  • 2 in length for versatile application.
  • Smooth 4 microinch finish or better for precise measurements.
  • Certificate of accuracy with NIST traceability included.