Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.5005 in, 2 in
Catalog No. 5000535376
Encompass
This Class X Plus Pin Gage is a precision instrument designed for accurate measurement and calibration. It features a 0.5005 inch diameter and a 2 inch length, with a 4 micro finish for enhanced durability and performance. Made from high-quality tool steel, it is ideal for calibrating measuring equipment or directly gaging parts.
  • 0.5005 inch diameter
  • 2 inch length
  • 0.00004 inch tolerance
  • Constructed from tool steel
  • 4 micro finish
  • Suitable for calibration of measuring equipment
  • Can be used to gage parts directly
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-35376 Supplier MSC Supplier No. 88750054
This Class X Plus Pin Gage is a precision instrument designed for accurate measurement and calibration. It features a 0.5005 inch diameter and a 2 inch length, with a 4 micro finish for enhanced durability and performance. Made from high-quality tool steel, it is ideal for calibrating measuring equipment or directly gaging parts.
  • 0.5005 inch diameter
  • 2 inch length
  • 0.00004 inch tolerance
  • Constructed from tool steel
  • 4 micro finish
  • Suitable for calibration of measuring equipment
  • Can be used to gage parts directly
  • Includes certificate of accuracy with NIST traceability