Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.5050" Dia, 2" Long, 4 Micro Coated
Catalog No. 5000535377
Encompass
This Class X Plus Pin Gage is engineered for precise measurement applications, featuring a 0.5050 inch diameter and a 2 inch length with a 4 Micro coating for enhanced accuracy. It is ideal for calibrating measuring equipment or directly gaging parts, ensuring reliability in various technical settings.
  • Diameter of measurement: 0.5050 inch
  • Tolerance: 0.00004 inch
  • Made from tool steel
  • Suitable for calibration of measuring equipment or direct gaging of parts
  • Includes a certificate of accuracy with NIST traceability
  • Length: 2 inch
  • Features a 4 Micro coating
Catalog No. 50-005-35377 Supplier MSC Supplier No. 88750500
This Class X Plus Pin Gage is engineered for precise measurement applications, featuring a 0.5050 inch diameter and a 2 inch length with a 4 Micro coating for enhanced accuracy. It is ideal for calibrating measuring equipment or directly gaging parts, ensuring reliability in various technical settings.
  • Diameter of measurement: 0.5050 inch
  • Tolerance: 0.00004 inch
  • Made from tool steel
  • Suitable for calibration of measuring equipment or direct gaging of parts
  • Includes a certificate of accuracy with NIST traceability
  • Length: 2 inch
  • Features a 4 Micro coating