Learn More
MSC Vermont Gage Class X Plus Pin Gage, 0.5700 In Diameter, 2 In Long
Catalog No. 5000535428
Encompass
This Class X Plus Pin Gage features a 0.5700 in diameter and a 2 in length. It is 4 Micro Coated and includes a Certificate of Accuracy with NIST Traceability, making it suitable for calibrating measuring equipment or directly gaging parts.
  • Class X Plus Pin Gage
  • 0.5700 in diameter
  • 2 in long
  • 4 micro coated
  • Includes certificate of accuracy with NIST traceability
  • Made of tool steel
  • Suitable for calibration of measuring equipment or direct part gaging
Catalog No. 50-005-35428 Supplier MSC Supplier No. 88757000
This Class X Plus Pin Gage features a 0.5700 in diameter and a 2 in length. It is 4 Micro Coated and includes a Certificate of Accuracy with NIST Traceability, making it suitable for calibrating measuring equipment or directly gaging parts.
  • Class X Plus Pin Gage
  • 0.5700 in diameter
  • 2 in long
  • 4 micro coated
  • Includes certificate of accuracy with NIST traceability
  • Made of tool steel
  • Suitable for calibration of measuring equipment or direct part gaging