Learn More
MSC Vermont Gage Class ZZ Go Pin Gage, 0.6030 inch, Tool Steel, 2 inch
Catalog No. 5000530542
Encompass
This Class ZZ Go Pin Gage is designed for precise measurement and inspection tasks in various industrial applications. It features a tight tolerance for accuracy and is constructed from durable tool steel.
  • Class ZZ tolerance ensures high accuracy
  • Constructed from durable tool steel with a hardness of 60/65 Rc
  • 2 inch length for versatile use
  • Surface finish of 10 microinch or better
  • Roundness maintained within 0.0001 inch
  • Marked with diameter and tolerance direction for easy identification
  • Includes a certificate of accuracy
Catalog No. 50-005-30542 Supplier MSC Supplier No. 88960307
This Class ZZ Go Pin Gage is designed for precise measurement and inspection tasks in various industrial applications. It features a tight tolerance for accuracy and is constructed from durable tool steel.
  • Class ZZ tolerance ensures high accuracy
  • Constructed from durable tool steel with a hardness of 60/65 Rc
  • 2 inch length for versatile use
  • Surface finish of 10 microinch or better
  • Roundness maintained within 0.0001 inch
  • Marked with diameter and tolerance direction for easy identification
  • Includes a certificate of accuracy