Learn More
MSC Vermont Gage Class ZZ Minus Gage, 0.1875 in, 2 in
Catalog No. 5000530473
Encompass
This Class ZZ Minus Plug & Pin Gage features a 0.1875 in diameter and 2 in overall length. Manufactured from steel with a 10 micro finish, it is designed for precise measurement applications such as setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • 0.1875 in diameter
  • 2 in overall length
  • 0.000200 in tolerance
  • Constructed from steel
  • Features a 10 micro finish
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-30473 Supplier MSC Supplier No. 89018758
This Class ZZ Minus Plug & Pin Gage features a 0.1875 in diameter and 2 in overall length. Manufactured from steel with a 10 micro finish, it is designed for precise measurement applications such as setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • 0.1875 in diameter
  • 2 in overall length
  • 0.000200 in tolerance
  • Constructed from steel
  • Features a 10 micro finish
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Includes certificate of accuracy with NIST traceability