Learn More
MSC Vermont Gage Class ZZ Minus Gage, 0.47 mm Dia, 2 in OAL
Catalog No. 5000533919
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection in various applications. Crafted from steel, it features a 0.47 mm diameter and a 2 inch overall length, providing accurate measurements with a tolerance of 0.00508 mm.
  • Constructed from durable steel.
  • Features a 10 micro coating and finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Can be used to set micrometers.
  • Suitable for NoGo inspection.
  • Applicable for use with verniers and other measuring devices.
Catalog No. 50-005-33919 Supplier MSC Supplier No. 40427924
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection in various applications. Crafted from steel, it features a 0.47 mm diameter and a 2 inch overall length, providing accurate measurements with a tolerance of 0.00508 mm.
  • Constructed from durable steel.
  • Features a 10 micro coating and finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Can be used to set micrometers.
  • Suitable for NoGo inspection.
  • Applicable for use with verniers and other measuring devices.