Learn More
MSC Vermont Gage Class ZZ Minus Gage, 0.7020 in, 2 in, Steel
Catalog No. 5000531652
Encompass
This steel plug and pin gage is designed for precise measurement and inspection tasks. With a 0.7020 in diameter and 2 in overall length, it is suitable for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Tolerance of 0.000200 in.
  • Features a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers.
  • Suitable for NoGo inspection.
  • Can be used with verniers and other measuring devices.
Catalog No. 50-005-31652 Supplier MSC Supplier No. 89070205
This steel plug and pin gage is designed for precise measurement and inspection tasks. With a 0.7020 in diameter and 2 in overall length, it is suitable for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Tolerance of 0.000200 in.
  • Features a 10 micro coating/finish.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers.
  • Suitable for NoGo inspection.
  • Can be used with verniers and other measuring devices.