Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.0150 in, 2 in
Catalog No. 5000537215
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Constructed from durable steel with a 10 micro finish, it features a 0.0150 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices. Each gage includes a certificate of accuracy with NIST traceability.
  • 0.0150 inch diameter for precise measurements.
  • 2 inch overall length.
  • Tolerance of 0.000200 inch ensures accuracy.
  • Made from durable steel.
  • Features a 10 micro coating/finish.
  • Suitable for setting micrometers, NoGo inspection, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-37215 Supplier MSC Supplier No. 89001507
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Constructed from durable steel with a 10 micro finish, it features a 0.0150 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices. Each gage includes a certificate of accuracy with NIST traceability.
  • 0.0150 inch diameter for precise measurements.
  • 2 inch overall length.
  • Tolerance of 0.000200 inch ensures accuracy.
  • Made from durable steel.
  • Features a 10 micro coating/finish.
  • Suitable for setting micrometers, NoGo inspection, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.