Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.0305 in Diameter, 2 in Length, Steel
Catalog No. 5000515418
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for quick and accurate measurements in various industrial applications. Made from durable steel, it features a precise 0.0305 inch diameter and an overall length of 2 inches, ensuring reliable performance for gaging slot widths, checking locations, and measuring hole sizes.
  • Measures hole sizes, distances between holes, and slot widths.
  • Checks locations and can be used for Go/NoGo gaging.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Constructed from steel for durability.
  • Features a 10 microinch finish for improved performance.
  • Provides an accuracy of 0.0002 "
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-15418 Supplier MSC Supplier No. 89003057
This Class ZZ Minus Plug & Pin Gage is designed for quick and accurate measurements in various industrial applications. Made from durable steel, it features a precise 0.0305 inch diameter and an overall length of 2 inches, ensuring reliable performance for gaging slot widths, checking locations, and measuring hole sizes.
  • Measures hole sizes, distances between holes, and slot widths.
  • Checks locations and can be used for Go/NoGo gaging.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Constructed from steel for durability.
  • Features a 10 microinch finish for improved performance.
  • Provides an accuracy of 0.0002 "
  • Includes a certificate of accuracy with NIST traceability.