Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.0395 in Diameter, 2 in Length, 10 Microfinish
Catalog No. 5000531541
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection applications. Crafted from durable steel, it features a 0.0395 inch diameter and a 2 inch overall length, with a 10 microfinish for accuracy. It is ideal for NoGo inspection and calibrating various measuring devices.
  • Application for NoGo inspection, setting micrometers, verniers, and other measuring devices.
  • Constructed from steel.
  • Features a 10 microfinish.
  • Includes a certificate of accuracy with NIST traceability.
  • Diameter of measurement is 0.0395 "
  • Overall length is 2 "
  • Tolerance of 0.000200 "
Catalog No. 50-005-31541 Supplier MSC Supplier No. 89003958
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection applications. Crafted from durable steel, it features a 0.0395 inch diameter and a 2 inch overall length, with a 10 microfinish for accuracy. It is ideal for NoGo inspection and calibrating various measuring devices.
  • Application for NoGo inspection, setting micrometers, verniers, and other measuring devices.
  • Constructed from steel.
  • Features a 10 microfinish.
  • Includes a certificate of accuracy with NIST traceability.
  • Diameter of measurement is 0.0395 "
  • Overall length is 2 "
  • Tolerance of 0.000200 "