Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.0475 in Dia, 2 in OAL, 10 Micro
Catalog No. 5000530884
Encompass
This Class ZZ Minus Plug & Pin Gage from Vermont Gage is designed for precise measurement applications. It features a 0.0475-in diameter and a 2-in overall length, with a 10 Micro finish. This gage is made from steel and is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. A Certificate of Accuracy with NIST Traceability is included to ensure its precision.
  • Made from steel
  • Suitable for setting micrometers
  • Useful for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-30884 Supplier MSC Supplier No. 71873160
This Class ZZ Minus Plug & Pin Gage from Vermont Gage is designed for precise measurement applications. It features a 0.0475-in diameter and a 2-in overall length, with a 10 Micro finish. This gage is made from steel and is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. A Certificate of Accuracy with NIST Traceability is included to ensure its precision.
  • Made from steel
  • Suitable for setting micrometers
  • Useful for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability