Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.0615 in Dia, 2 in OAL, Steel
Catalog No. 5000534705
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for various inspection and calibration tasks. It features a 0.0615 inch diameter and a 2 inch overall length, crafted from durable steel with a black oxide finish for enhanced protection.
  • Crafted from durable steel
  • Features a black oxide coating for enhanced protection
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices
  • Manufactured in the United States
  • Tolerance of 0.000200 inch ensures high precision
Catalog No. 50-005-34705 Supplier MSC Supplier No. 74511254
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for various inspection and calibration tasks. It features a 0.0615 inch diameter and a 2 inch overall length, crafted from durable steel with a black oxide finish for enhanced protection.
  • Crafted from durable steel
  • Features a black oxide coating for enhanced protection
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices
  • Manufactured in the United States
  • Tolerance of 0.000200 inch ensures high precision