Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.0810 in, 2 in
Catalog No. 5000528939
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. It features a 0.0810 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, no-go inspection, and use with verniers and other measuring devices. Constructed from steel with a black oxide finish, it offers durability and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for setting micrometers, no-go inspection, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Made from durable steel.
  • Features a protective black oxide finish.
Catalog No. 50-005-28939 Supplier MSC Supplier No. 79208336
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. It features a 0.0810 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, no-go inspection, and use with verniers and other measuring devices. Constructed from steel with a black oxide finish, it offers durability and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for setting micrometers, no-go inspection, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Made from durable steel.
  • Features a protective black oxide finish.