Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.1235 in Dia, 2 in OAL
Catalog No. 5000515634
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications, including setting micrometers, verniers, and for NoGo inspection. It features a diameter of 0.1235 inches and an overall length of 2 inches, providing accurate and reliable performance.
  • Made from steel for durability
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Tolerance of 0.000200 inches
Catalog No. 50-005-15634 Supplier MSC Supplier No. 89012355
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications, including setting micrometers, verniers, and for NoGo inspection. It features a diameter of 0.1235 inches and an overall length of 2 inches, providing accurate and reliable performance.
  • Made from steel for durability
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Tolerance of 0.000200 inches