Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.1280 in Diameter, 2 in OAL, Steel
Catalog No. 5000515366
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. Made of steel with a 10 micro finish, it features a 0.1280 in diameter and a 2 in overall length, making it ideal for setting micrometers and for NoGo inspection.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection.
  • Constructed from durable steel.
  • Features a 10 micro finish.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-15366 Supplier MSC Supplier No. 89012801
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. Made of steel with a 10 micro finish, it features a 0.1280 in diameter and a 2 in overall length, making it ideal for setting micrometers and for NoGo inspection.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection.
  • Constructed from durable steel.
  • Features a 10 micro finish.
  • Includes a certificate of accuracy with NIST traceability.