Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.1370 in Dia, 2 in OAL, Steel
Catalog No. 5000537218
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection applications. With a 0.1370 inch diameter and 2 inch overall length, it is suitable for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Made of steel
  • 10 micro finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Manufactured in the United States
  • Tolerance of 0.000200 in
Catalog No. 50-005-37218 Supplier MSC Supplier No. 89013700
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection applications. With a 0.1370 inch diameter and 2 inch overall length, it is suitable for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Made of steel
  • 10 micro finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Manufactured in the United States
  • Tolerance of 0.000200 in