Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.1370 in Dia, 2 in OAL
Catalog No. 5000511407
Encompass
This Class ZZ minus plug and pin gage is designed for accurate measurement and inspection tasks. It features a 0.1370 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, NoGo inspection, and other measuring devices.
  • Made from durable steel.
  • Features a black oxide finish.
  • Measurement diameter of 0.1370 "
  • Overall length of 2 inches.
  • Tolerance of 0.000200 "
  • Ideal for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured in the United States.
Catalog No. 50-005-11407 Supplier MSC Supplier No. 02711216
This Class ZZ minus plug and pin gage is designed for accurate measurement and inspection tasks. It features a 0.1370 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, NoGo inspection, and other measuring devices.
  • Made from durable steel.
  • Features a black oxide finish.
  • Measurement diameter of 0.1370 "
  • Overall length of 2 inches.
  • Tolerance of 0.000200 "
  • Ideal for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured in the United States.