Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.1830 in Dia, 2 in OAL, Steel
Catalog No. 5000515766
Encompass
This Class ZZ Minus Plug & Pin Gage features a diameter of 0.1830 in and an overall length of 2 in. Constructed from steel with a 0.000200 in tolerance, it is designed for precise measurement applications.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability
  • Features a 10 Micro coating/finish
Catalog No. 50-005-15766 Supplier MSC Supplier No. 89018303
This Class ZZ Minus Plug & Pin Gage features a diameter of 0.1830 in and an overall length of 2 in. Constructed from steel with a 0.000200 in tolerance, it is designed for precise measurement applications.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a Certificate of Accuracy with NIST Traceability
  • Features a 10 Micro coating/finish