Learn More
MSC Vermont Gage Class ZZ Minus Plug & Pin Gage, 0.2050 in, 2 in
Catalog No. 5000531164
Encompass
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision instrument designed for accurate measurement and inspection. It features a 0.2050 inch diameter and a 2 inch overall length, made from durable steel with a 10 Micro finish. This gage is supplied with a Certificate of Accuracy that offers NIST Traceability, ensuring reliable performance in various applications.
  • Designed for high-speed applications.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Made of steel for durability.
  • Features a 10 Micro finish.
Catalog No. 50-005-31164 Supplier MSC Supplier No. 89020507
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision instrument designed for accurate measurement and inspection. It features a 0.2050 inch diameter and a 2 inch overall length, made from durable steel with a 10 Micro finish. This gage is supplied with a Certificate of Accuracy that offers NIST Traceability, ensuring reliable performance in various applications.
  • Designed for high-speed applications.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Made of steel for durability.
  • Features a 10 Micro finish.